Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Iodophenyl)piperazine hydrochloride, 95%

Chemical Identifiers

Chemical Name or Material 1-(4-Iodophenyl)piperazine hydrochloride
CAS 624726-35-8
Formula Weight 324.59
MDL Number MFCD04119724
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5529903

 
alfa aesar™
H5529903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5529906

 
alfa aesar™
H5529906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 324.59
Melting Point 177° to 181°C
Chemical Name or Material 1-(4-Iodophenyl)piperazine hydrochloride
Physical Form Solid
Assay 95%
MDL Number MFCD04119724
CAS 624726-35-8
Quantity 1g
Sensitivity Light Sensitive