missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Iodophenyl)-2-methyl-1H-imidazole, 97%, Maybridge

Chemical Identifiers

CAS 179420-75-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H9IN2
Formula Weight 284.1
MDL Number MFCD09261230
Chemical Name or Material 1-(4-Iodophenyl)-2-methyl-1H-imidazole
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC61411DA SDS Fisher Scientific Company
CC61411DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC61411CB SDS Fisher Scientific Company
CC61411CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

179420-75-8
100.0
C10H9IN2
MFCD09261230
97%
96.5
97%
284.1
1-(4-Iodophenyl)-2-methyl-1H-imidazole

Specifications

284.1
100.0
97%
MFCD09261230
Amber Glass Bottle
97%
1g
179420-75-8
C10H9IN2
96.5
1-(4-Iodophenyl)-2-methyl-1H-imidazole

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning