missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Iodobenzyl)pyrrolidine, ≥97%, Maybridge

Chemical Identifiers

CAS 858676-60-5
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H14IN
Formula Weight 287.14
MDL Number MFCD07775752
Chemical Name or Material 1-(4-Iodobenzyl)pyrrolidine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC55611DA SDS Fisher Scientific Company
CC55611DA
Amber glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC55611DE Fisher Scientific Company
CC55611DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

858676-60-5
100.0
C11H14IN
MFCD07775752
97%
96.5
97%
287.14
1-(4-Iodobenzyl)pyrrolidine

Specifications

287.14
100.0
C11H14IN
96.5
MFCD07775752
97%
1g
97%
858676-60-5
Amber glass bottle
1-(4-Iodobenzyl)pyrrolidine

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger