missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Hydrazinobenzyl)-1H-1,2,4-triazole Dihydrochloride, TCI America™

$132.92 - $453.77

Chemical Identifiers

CAS 212248-62-9
Molecular Formula C9H11N5
Molecular Weight (g/mol) 189.222
MDL Number MFCD06795497
InChI Key FUJKUTJPEGEMHG-UHFFFAOYSA-N
Synonym 4-[(1,2,4-Triazol-1-yl)methyl]phenylhydrazine Dihydrochloride
PubChem CID 7472049
IUPAC Name [4-(1,2,4-triazol-1-ylmethyl)phenyl]hydrazine
SMILES C1=CC(=CC=C1CN2C=NC=N2)NN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H1467200MG View Documents TCI America
H1467200MG
200mg Each for $132.92
 
H14671G View Documents TCI America
H14671G
1g Each for $453.77
 
Specifications

Chemical Identifiers

212248-62-9
189.222
FUJKUTJPEGEMHG-UHFFFAOYSA-N
7472049
C1=CC(=CC=C1CN2C=NC=N2)NN
C9H11N5
MFCD06795497
4-[(1,2,4-Triazol-1-yl)methyl]phenylhydrazine Dihydrochloride
[4-(1,2,4-triazol-1-ylmethyl)phenyl]hydrazine

Specifications

Undesignated
212248-62-9
MFCD06795497
FUJKUTJPEGEMHG-UHFFFAOYSA-N
[4-(1,2,4-triazol-1-ylmethyl)phenyl]hydrazine
7472049
Crystal-Powder at 20°C
1-(4-Hydrazinobenzyl)-1H-1,2,4-triazole Dihydrochloride
200mg
C9H11N5
4-[(1,2,4-Triazol-1-yl)methyl]phenylhydrazine Dihydrochloride
C1=CC(=CC=C1CN2C=NC=N2)NN
189.222
262.14
≥97.0% (HPLC,T)
Safety and Handling

Safety and Handling

TSCA : No