Please login to your online account to display your discounted pricing

1,4-Hexadiene, Mixture of cis and trans 99%, Acros Organics


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  156550100

 View more versions of this product

Catalog No. AC156550100

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 82.13
Physical Form Liquid
Assay ≥98.5%
Quantity 10g
Color Clear colorless
Beilstein 01, 253
TSCA TSCA
Density 0.7100g/mL
EINECS Number 209-756-8
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
CAS Min % 98.5
Refractive Index 1.4100 to 1.4120 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Name Note 99%
Linear Formula CH3CH=CHCH2CH=CH2
MDL Number MFCD00009298
Assay Percent Range 99%
Chemical Name or Material 1, 4-Hexadiene
CAS 592-45-0
Molecular Formula C6H10

Specifications

Chemical Name or Material 1, 4-Hexadiene
Name Note 99%
CAS 592-45-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H10
Linear Formula CH3CH=CHCH2CH=CH2
Formula Weight 82.13
MDL Number MFCD00009298
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents