Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Fluorophenyl)pyrrole, 98%

$57.57 - $206.43

Chemical Identifiers

Chemical Name or Material 1-(4-Fluorophenyl)pyrrole
CAS 81329-31-9
Molecular Formula C10H8FN
Formula Weight 161.18
MDL Number MFCD00047066
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1395703

  SDS
alfa aesar™
A1395703
1g Each for $57.57

AAA1395706

  SDS
alfa aesar™
A1395706
5g Each for $206.43
Description & Specifications

Specifications

Formula Weight 161.18
Chemical Name or Material 1-(4-Fluorophenyl)pyrrole
Molecular Formula C10H8FN
Melting Point 50° to 52°C
CAS 81329-31-9
MDL Number MFCD00047066
Assay 98%
Quantity 1g