missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Fluorophenyl)piperazine dihydrochloride, 98%

$42.09 - $425.62

Chemical Identifiers

CAS 64090-19-3
Molecular Formula C10H15Cl2FN2
Molecular Weight (g/mol) 253.142
MDL Number MFCD00012765
InChI Key DZQVAQAZQDURKX-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl piperazine dihydrochloride, 1-4-fluorophenyl piperazinedihydrochloride, 1-4-fluorophenyl piperazine 2hcl, piperazine, 1-4-fluorophenyl-, dihydrochloride, 1-4-fluorophenyl piperazine;dihydrochloride, para-fluorophenylpiperazine dihydrochloride, zlchem 337, pubchem15283, acmc-209nk7, ksc352o8p
PubChem CID 16211916
IUPAC Name 1-(4-fluorophenyl)piperazine;dihydrochloride
SMILES C1CN(CCN1)C2=CC=C(C=C2)F.Cl.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1709714 View Documents Alfa Aesar™
L1709714
25g Each for $42.09
 
AAL1709722 View Documents Alfa Aesar™
L1709722
100g Each for $104.24
 
AAL1709736 View Documents Alfa Aesar™
L1709736
500g Each for $425.62
 
Specifications

Chemical Identifiers

64090-19-3
253.142
DZQVAQAZQDURKX-UHFFFAOYSA-N
16211916
C1CN(CCN1)C2=CC=C(C=C2)F.Cl.Cl
C10H15Cl2FN2
MFCD00012765
1-4-fluorophenyl piperazine dihydrochloride, 1-4-fluorophenyl piperazinedihydrochloride, 1-4-fluorophenyl piperazine 2hcl, piperazine, 1-4-fluorophenyl-, dihydrochloride, 1-4-fluorophenyl piperazine;dihydrochloride, para-fluorophenylpiperazine dihydrochloride, zlchem 337, pubchem15283, acmc-209nk7, ksc352o8p
1-(4-fluorophenyl)piperazine;dihydrochloride

Specifications

~234°C
64090-19-3
MFCD00012765
4826840
DZQVAQAZQDURKX-UHFFFAOYSA-N
1-(4-fluorophenyl)piperazine;dihydrochloride
16211916
98%
25g
C10H15Cl2FN2
UN2811
1-4-fluorophenyl piperazine dihydrochloride, 1-4-fluorophenyl piperazinedihydrochloride, 1-4-fluorophenyl piperazine 2hcl, piperazine, 1-4-fluorophenyl-, dihydrochloride, 1-4-fluorophenyl piperazine;dihydrochloride, para-fluorophenylpiperazine dihydrochloride, zlchem 337, pubchem15283, acmc-209nk7, ksc352o8p
C1CN(CCN1)C2=CC=C(C=C2)F.Cl.Cl
253.142
253.15
1-(4-Fluorophenyl)piperazine dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.