missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Fluorophenyl)piperazine, 98%, Thermo Scientific™

$45.20 - $45.20

Chemical Identifiers

CAS 2252-63-3
Molecular Formula C10H13FN2
Molecular Weight (g/mol) 180.22
InChI Key AVJKDKWRVSSJPK-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl piperazine, 1-p-fluorophenyl piperazine, 4-fluoro-phenylpiperazine, 4-fluorophenyl piperazine, 4-fluorophenylpiperazine, n-4-fluorophenyl piperazine, para-fluorophenylpiperazine, pfpp, piperazine, 1-4-fluorophenyl, unii-vml61be244
PubChem CID 75260
IUPAC Name 1-(4-fluorophenyl)piperazine
SMILES C1CN(CCN1)C2=CC=C(C=C2)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC184540100
SDS View Documents Promotion Details
Thermo Fisher Scientific
184540100
10g Glass bottle
Each for $45.20
 
Description

Description

Specifications

Chemical Identifiers

2252-63-3
180.22
1-4-fluorophenyl piperazine, 1-p-fluorophenyl piperazine, 4-fluoro-phenylpiperazine, 4-fluorophenyl piperazine, 4-fluorophenylpiperazine, n-4-fluorophenyl piperazine, para-fluorophenylpiperazine, pfpp, piperazine, 1-4-fluorophenyl, unii-vml61be244
1-(4-fluorophenyl)piperazine
C10H13FN2
AVJKDKWRVSSJPK-UHFFFAOYSA-N
75260
C1CN(CCN1)C2=CC=C(C=C2)F

Specifications

2252-63-3
100.0
>110°C
C10H13FN2
Glass bottle
10g
AVJKDKWRVSSJPK-UHFFFAOYSA-N
1-(4-fluorophenyl)piperazine
75260
Liquid After Melting
1-(4-Fluorophenyl)piperazine
97.5
Yellow
97.5% min. (GC)
Authentic
1.5585 to 1.5605
1-4-fluorophenyl piperazine, 1-p-fluorophenyl piperazine, 4-fluoro-phenylpiperazine, 4-fluorophenyl piperazine, 4-fluorophenylpiperazine, n-4-fluorophenyl piperazine, para-fluorophenylpiperazine, pfpp, piperazine, 1-4-fluorophenyl, unii-vml61be244
C1CN(CCN1)C2=CC=C(C=C2)F
180.22
180.22
98%
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 218-846-6

SDS
Product Certifications

RUO – Research Use Only