missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Fluorophenyl)piperazine, 98%

Manufacturer:  Alfa Aesar™ A1454106

 View more versions of this product

Catalog No. AAA1454106



Description

Description

Applications
1-(4-Fluorophenyl)piperazine was used in the synthesis of N,N-disubstituted piperazine. It is a major metabolite of niaparazine, a sedative, a hypnotic drug.

Solubility
Insoluble in water.

Notes
Air sensitive. Store under inert gas. Store away from air, oxidizing agents.
Specifications

Specifications

1-(4-Fluorophenyl)piperazine
120°C (0.1mmHg)
5g
C10H13FN2
UN2923
1-4-fluorophenyl piperazine, 4-fluorophenylpiperazine, pfpp, 4-fluoro-phenylpiperazine, n-4-fluorophenyl piperazine, para-fluorophenylpiperazine, unii-vml61be244, 1-p-fluorophenyl piperazine, piperazine, 1-4-fluorophenyl, 4-fluorophenyl piperazine
C1CN(CCN1)C2=CC=C(C=C2)F
180.226
180.23
>110°C (230°F)
30°C to 33°C
2252-63-3
MFCD00005960
610723
AVJKDKWRVSSJPK-UHFFFAOYSA-N
1-(4-fluorophenyl)piperazine
75260
98%

Chemical Identifiers

2252-63-3
180.226
AVJKDKWRVSSJPK-UHFFFAOYSA-N
75260
C1CN(CCN1)C2=CC=C(C=C2)F
C10H13FN2
MFCD00005960
1-4-fluorophenyl piperazine, 4-fluorophenylpiperazine, pfpp, 4-fluoro-phenylpiperazine, n-4-fluorophenyl piperazine, para-fluorophenylpiperazine, unii-vml61be244, 1-p-fluorophenyl piperazine, piperazine, 1-4-fluorophenyl, 4-fluorophenyl piperazine
1-(4-fluorophenyl)piperazine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

Documents