Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Fluorophenyl)homopiperazine dihydrochloride, 98%

$672.30 - $672.30

Chemical Identifiers

Chemical Name or Material 1-(4-Fluorophenyl)homopiperazine dihydrochloride
CAS 263409-96-7
Molecular Formula C11H15FN2·2HCl
Formula Weight 267.17g/mol
MDL Number MFCD08436121
Synonym 4-FPHP·2HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51701ME

 
alfa aesar™
H51701-ME
Each for $672.30
Description & Specifications

Specifications

Melting Point 219° to 223°C
Quantity 500mg
Chemical Name or Material 1-(4-Fluorophenyl)homopiperazine dihydrochloride
CAS 263409-96-7
Assay 98%
Molecular Formula C11H15FN2·2HCl
Formula Weight 267.17g/mol
MDL Number MFCD08436121
Synonym 4-FPHP·2HCl
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.