Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-1-(4-Fluorophenyl)ethylamine, 97%

Chemical Identifiers

Chemical Name or Material (+/-)-1-(4-Fluorophenyl)ethylamine
CAS 403-40-7
Molecular Formula C8H10FN
Formula Weight 139.17
MDL Number MFCD00041323
UN Number UN2735
Beilstein 2802417
Synonym (+/-)-4-Fluoro-alpha-methylbenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0457503

  SDS
alfa aesar™
L0457503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0457506

  SDS
alfa aesar™
L0457506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 139.17
CAS 403-40-7
Molecular Formula C8H10FN
Synonym (+/-)-4-Fluoro-alpha-methylbenzylamine
UN Number UN2735
Assay 97%
Health Hazard 2 GHS H Statement
H314-H227
Causes severe skin burns and eye damage.
Combustible liquid.
pH 10
Quantity 1g
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 68°C (154°F)
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Beilstein 2802417
Odor Amine-like
Boiling Point 76°C (22mmHg)
MDL Number MFCD00041323
Chemical Name or Material (+/-)-1-(4-Fluorophenyl)ethylamine
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H227
Causes severe skin burns and eye damage.
Combustible liquid.