Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-1-(4-Fluorophenyl)ethyl isothiocyanate, 97%

Chemical Identifiers

Chemical Name or Material (+/-)-1-(4-Fluorophenyl)ethyl isothiocyanate
CAS 182565-27-1
Molecular Formula C9H8FNS
Formula Weight 181.23
MDL Number MFCD00041105
UN Number UN2922
Beilstein 6123780
Synonym 4-Fluoro-alpha-methylbenzyl isothiocyanate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1212303

  SDS
alfa aesar™
L1212303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.23
Chemical Name or Material (+/-)-1-(4-Fluorophenyl)ethyl isothiocyanate
MDL Number MFCD00041105
Assay 97%
Molecular Formula C9H8FNS
CAS 182565-27-1
Beilstein 6123780
Synonym 4-Fluoro-alpha-methylbenzyl isothiocyanate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
UN Number UN2922
Boiling Point 109° to 110°C (0.6mmHg)
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.