Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Fluorophenyl)ethanol, 98%

$510.80 - $510.80

Chemical Identifiers

Chemical Name or Material 1-(4-Fluorophenyl)ethanol
CAS 403-41-8
Molecular Formula C8H9FO
Formula Weight 140.16
MDL Number MFCD00004515
Beilstein 2042025
Synonym 4-Fluorophenyl methyl carbinol; 4-Fluoro-alpha-methylbenzyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2209722

  SDS
alfa aesar™
B2209722
100g Each for $510.80
Description & Specifications

Specifications

Formula Weight 140.16
Recommended Storage 2° to 8°C
Synonym 4-Fluorophenyl methyl carbinol; 4-Fluoro-alpha-methylbenzyl alcohol
Assay 98%
Molecular Formula C8H9FO
Beilstein 2042025
MDL Number MFCD00004515
Boiling Point 90° to 92°C (7mmHg)
Flash Point 91°C (196°F)
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
CAS 403-41-8
Chemical Name or Material 1-(4-Fluorophenyl)ethanol
Health Hazard 1 Warning
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

2° to 8°C