Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Fluorophenyl)biguanide hydrochloride, 98%

$43.08 - $150.68

Chemical Identifiers

Chemical Name or Material 1-(4-Fluorophenyl)biguanide hydrochloride
CAS 16018-83-0
Molecular Formula C8H10FN5·HCl
Formula Weight 231.66
MDL Number MFCD00051986
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0733003

 
alfa aesar™
L0733003
1g Each for $43.08

AAL0733006

 
alfa aesar™
L0733006
5g Each for $150.68
Description & Specifications

Specifications

Formula Weight 231.66
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 1-(4-Fluorophenyl)biguanide hydrochloride
Melting Point 212° to 216°C
MDL Number MFCD00051986
CAS 16018-83-0
Molecular Formula C8H10FN5·HCl
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Quantity 1g
RTECS Number DU1850000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.