Please login to your online account to display your discounted pricing

1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride, 97%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CC00119DE

 View more versions of this product

Catalog No. CC00119DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 238.65
Quantity 5g
Assay 97%
CAS Max % 100.0
Molecular Formula C11H8ClFN2O
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
MDL Number MFCD03659694
CAS 423768-49-4
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

Specifications

Chemical Name or Material 1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride
CAS 423768-49-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H8ClFN2O
Formula Weight 238.65
MDL Number MFCD03659694
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.