Please login to your online account to display your discounted pricing

[1-(4-Fluorophenyl)-5-methyl-1H-pyrazol-4-yl]methanol, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material [1-(4-Fluorophenyl)-5-methyl-1H-pyrazol-4-yl]methanol
CAS 465514-37-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H11FN2O
Formula Weight 206.22
MDL Number MFCD03644148
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00765CB

  SDS
fisher scientific company
MO00765CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 206.22
Quantity 250mg
Assay 97%
CAS Min % 97.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Range 97%
CAS 465514-37-8
Molecular Formula C11H11FN2O
MDL Number MFCD03644148
Chemical Name or Material [1-(4-Fluorophenyl)-5-methyl-1H-pyrazol-4-yl]methanol
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.