missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Fluorobenzyl)piperazine, 97%

$47.81 - $154.22

Chemical Identifiers

CAS 70931-28-1
Molecular Formula C11H15FN2
Molecular Weight (g/mol) 194.253
MDL Number MFCD01075234
InChI Key OOSZCNKVJAVHJI-UHFFFAOYSA-N
Synonym 1-4-fluorobenzyl piperazine, 1-4-fluorophenyl methyl piperazine, 1-4-fluorobenzyl-piperazine, piperazine,1-4-fluorophenyl methyl, 1-4-fluoro-benzyl-piperazine, 4-fluorobenzylpiperazine, piperazine, 1-4-fluorophenyl methyl, 4-fluorophenyl methyl piperazine, 4-fluorobenzyl-piperazine, acmc-1bl7k
PubChem CID 796563
IUPAC Name 1-[(4-fluorophenyl)methyl]piperazine
SMILES C1CN(CCN1)CC2=CC=C(C=C2)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6199306 View Documents Alfa Aesar™
H6199306
5g Each for $47.81
 
AAH6199314 View Documents Alfa Aesar™
H6199314
25g Each for $154.22
 
Specifications

Chemical Identifiers

70931-28-1
194.253
OOSZCNKVJAVHJI-UHFFFAOYSA-N
796563
C1CN(CCN1)CC2=CC=C(C=C2)F
C11H15FN2
MFCD01075234
1-4-fluorobenzyl piperazine, 1-4-fluorophenyl methyl piperazine, 1-4-fluorobenzyl-piperazine, piperazine,1-4-fluorophenyl methyl, 1-4-fluoro-benzyl-piperazine, 4-fluorobenzylpiperazine, piperazine, 1-4-fluorophenyl methyl, 4-fluorophenyl methyl piperazine, 4-fluorobenzyl-piperazine, acmc-1bl7k
1-[(4-fluorophenyl)methyl]piperazine

Specifications

63°C to 67°C
70931-28-1
MFCD01075234
1-4-fluorobenzyl piperazine, 1-4-fluorophenyl methyl piperazine, 1-4-fluorobenzyl-piperazine, piperazine,1-4-fluorophenyl methyl, 1-4-fluoro-benzyl-piperazine, 4-fluorobenzylpiperazine, piperazine, 1-4-fluorophenyl methyl, 4-fluorophenyl methyl piperazine, 4-fluorobenzyl-piperazine, acmc-1bl7k
C1CN(CCN1)CC2=CC=C(C=C2)F
194.253
194.25
1-(4-Fluorobenzyl)piperazine
5g
C11H15FN2
UN3259
OOSZCNKVJAVHJI-UHFFFAOYSA-N
1-[(4-fluorophenyl)methyl]piperazine
796563
97%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.