Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Fluoro-2-nitrophenyl)pyrrolidine, 97%

Chemical Identifiers

Chemical Name or Material 1-(4-Fluoro-2-nitrophenyl)pyrrolidine
CAS 778-56-3
Formula Weight 210.2
MDL Number MFCD00798372
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6320403

 
alfa aesar™
H6320403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6320406

 
alfa aesar™
H6320406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6320414

 
alfa aesar™
H6320414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.2
Chemical Name or Material 1-(4-Fluoro-2-nitrophenyl)pyrrolidine
Physical Form Solid
Assay 97%
MDL Number MFCD00798372
CAS 778-56-3
Quantity 1g