Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Fluoro-2-nitrophenyl)piperidine, 97%

Chemical Identifiers

Chemical Name or Material 1-(4-Fluoro-2-nitrophenyl)piperidine
CAS 719-70-0
Formula Weight 224.23
MDL Number MFCD00798370
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6314203

 
alfa aesar™
H6314203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6314206

 
alfa aesar™
H6314206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6314214

 
alfa aesar™
H6314214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.23
Chemical Name or Material 1-(4-Fluoro-2-nitrophenyl)piperidine
Physical Form Solid
Assay 97%
MDL Number MFCD00798370
CAS 719-70-0
Quantity 1g