missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(4-Fluoro-2-nitrophenyl)piperazine, 97%, Thermo Scientific™
$105.73 - $448.22
Chemical Identifiers
CAS | 243128-46-3 |
---|---|
Molecular Formula | C10H12FN3O2 |
Molecular Weight (g/mol) | 225.22 |
MDL Number | MFCD00798371 |
InChI Key | ZPYLKZRKXDCEQI-UHFFFAOYSA-N |
Synonym | 1-4-fluoro-2-nitrophenyl piperazine, 4-fluoro-2-nitrophenyl piperazine, piperazine, 1-4-fluoro-2-nitrophenyl, 1-4-fluoro-2-nitro-phenyl piperazine, 5-fluoro-2-piperazin-1-yl nitrobenzene |
PubChem CID | 2737435 |
IUPAC Name | 1-(4-fluoro-2-nitrophenyl)piperazine |
SMILES | [O-][N+](=O)C1=C(C=CC(F)=C1)N1CCNCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6313303
|
Thermo Scientific Chemicals
H6313303 |
1 g |
Each for $105.73
|
|
|||||
AAH6313306
|
Thermo Scientific Chemicals
H6313306 |
5 g |
Each for $448.22
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
243128-46-3 | |
225.22 | |
ZPYLKZRKXDCEQI-UHFFFAOYSA-N | |
2737435 | |
[O-][N+](=O)C1=C(C=CC(F)=C1)N1CCNCC1 |
C10H12FN3O2 | |
MFCD00798371 | |
1-4-fluoro-2-nitrophenyl piperazine, 4-fluoro-2-nitrophenyl piperazine, piperazine, 1-4-fluoro-2-nitrophenyl, 1-4-fluoro-2-nitro-phenyl piperazine, 5-fluoro-2-piperazin-1-yl nitrobenzene | |
1-(4-fluoro-2-nitrophenyl)piperazine |
Specifications
243128-46-3 | |
MFCD00798371 | |
1-4-fluoro-2-nitrophenyl piperazine, 4-fluoro-2-nitrophenyl piperazine, piperazine, 1-4-fluoro-2-nitrophenyl, 1-4-fluoro-2-nitro-phenyl piperazine, 5-fluoro-2-piperazin-1-yl nitrobenzene | |
[O-][N+](=O)C1=C(C=CC(F)=C1)N1CCNCC1 | |
225.22 | |
225.22 | |
Solid |
C10H12FN3O2 | |
1 g | |
ZPYLKZRKXDCEQI-UHFFFAOYSA-N | |
1-(4-fluoro-2-nitrophenyl)piperazine | |
2737435 | |
97% | |
1-(4-Fluoro-2-nitrophenyl)piperazine |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures