missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Fluoro-2-nitrophenyl)homopiperazine, 97%

$57.05 - $675.45

Chemical Identifiers

CAS 874814-95-6
Molecular Formula C11H14FN3O2
Molecular Weight (g/mol) 239.25
InChI Key HFLSWHSIIVSRPS-UHFFFAOYSA-N
Synonym 1-4-fluoro-2-nitrophenyl-1,4-diazepane, 1-4-fluoro-2-nitrophenyl-1,4-diazaperhydroepine, 1h-1,4-diazepine, 1-4-fluoro-2-nitrophenyl hexahydro
PubChem CID 18801299
IUPAC Name 1-(4-fluoro-2-nitrophenyl)-1,4-diazepane
SMILES C1CNCCN(C1)C2=C(C=C(C=C2)F)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63831MD View Documents Alfa Aesar™
H63831MD
250mg Each for $57.05
 
AAH6383103 View Documents Alfa Aesar™
H6383103
1g Each for $171.18
 
AAH6383106 View Documents Alfa Aesar™
H6383106
5g Each for $675.45
 
Specifications

Chemical Identifiers

874814-95-6
239.25
1-4-fluoro-2-nitrophenyl-1,4-diazepane, 1-4-fluoro-2-nitrophenyl-1,4-diazaperhydroepine, 1h-1,4-diazepine, 1-4-fluoro-2-nitrophenyl hexahydro
1-(4-fluoro-2-nitrophenyl)-1,4-diazepane
C11H14FN3O2
HFLSWHSIIVSRPS-UHFFFAOYSA-N
18801299
C1CNCCN(C1)C2=C(C=C(C=C2)F)[N+](=O)[O-]

Specifications

250mg
C11H14FN3O2
1-4-fluoro-2-nitrophenyl-1,4-diazepane, 1-4-fluoro-2-nitrophenyl-1,4-diazaperhydroepine, 1h-1,4-diazepine, 1-4-fluoro-2-nitrophenyl hexahydro
C1CNCCN(C1)C2=C(C=C(C=C2)F)[N+](=O)[O-]
239.25
239.24
97%
874814-95-6
UN3259
HFLSWHSIIVSRPS-UHFFFAOYSA-N
1-(4-fluoro-2-nitrophenyl)-1,4-diazepane
18801299
Solid
1-(4-Fluoro-2-nitrophenyl)homopiperazine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.