missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Fluoro-2-nitrophenyl)-4-piperidone, 97%

Chemical Identifiers

Formula Weight 238.22
Chemical Name or Material 1-(4-Fluoro-2-nitrophenyl)-4-piperidone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6388203 Alfa Aesar™
H6388203
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6388206 Alfa Aesar™
H6388206
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH63882MD Alfa Aesar™
H63882MD
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

238.22
1-(4-Fluoro-2-nitrophenyl)-4-piperidone

Specifications

1g
238.22
97%
Solid
1-(4-Fluoro-2-nitrophenyl)-4-piperidone