Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Fluoro-2-nitrophenyl)-4-methylpiperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-(4-Fluoro-2-nitrophenyl)-4-methylpiperazine
CAS 432531-43-6
Formula Weight 239.25
MDL Number MFCD03144445
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6319803

 
alfa aesar™
H6319803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6319806

 
alfa aesar™
H6319806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63198MD

 
alfa aesar™
H63198MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 239.25
Chemical Name or Material 1-(4-Fluoro-2-nitrophenyl)-4-methylpiperazine
Physical Form Solid
Assay 97%
MDL Number MFCD03144445
CAS 432531-43-6
Quantity 1g