Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene, 99+%

$290.92 - $290.92

Chemical Identifiers

Chemical Name or Material 1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene
CAS 95480-29-8
Molecular Formula C21H24O
Formula Weight 292.42
MDL Number MFCD01218020
Synonym 1-(4-Ethoxyphenyl)-2-(4-n-pentylphenyl)acetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2747609

  SDS
alfa aesar™
H2747609
10g Each for $290.92
Description & Specifications

Specifications

Formula Weight 292.42
Melting Point 62°C
Chemical Name or Material 1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene
Assay ≥99%
Molecular Formula C21H24O
MDL Number MFCD01218020
CAS 95480-29-8
Synonym 1-(4-Ethoxyphenyl)-2-(4-n-pentylphenyl)acetylene
Boiling Point 178° to 180°C (0.5mmHg)
Quantity 10g