Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Ethoxyphenyl)ethanol, 98%

$45.26 - $504.88

Chemical Identifiers

Chemical Name or Material 1-(4-Ethoxyphenyl)ethanol
CAS 52067-36-4
Molecular Formula C10H14O2
Formula Weight 166.22
MDL Number MFCD08062382
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3005203

 
alfa aesar™
H3005203
1g Each for $45.26

AAH3005206

 
alfa aesar™
H3005206
5g Each for $150.85

AAH3005214

 
alfa aesar™
H3005214
25g Each for $504.88
Description & Specifications

Specifications

Formula Weight 166.22
Chemical Name or Material 1-(4-Ethoxyphenyl)ethanol
Assay 98%
Molecular Formula C10H14O2
MDL Number MFCD08062382
CAS 52067-36-4
Melting Point 46° to 47°C
Quantity 1g