Please login to your online account to display your discounted pricing

1,4-Dithiothreitol, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

$402.25 - $2,867.45

Chemical Identifiers

Chemical Name or Material 1,4-Dithiothreitol
CAS 3483-12-3
Purity Grade Ultrapure Bioreagent
Molecular Formula HSCH2(CHOH)2CH2SH
Formula Weight 154.25
Synonym DTT; Cleland's Reagent
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002014

 
avantor performance
F78001
POLYSTORMOR 5g Each for $402.25

02002015

 
avantor performance
F78003
Poly Bottle 100g Each for $2,867.45

02002018

 
avantor performance
F78002
POLYSTORMOR 25g Each for $887.66
Description & Specifications

Specifications

Formula Weight 154.25
Chemical Name or Material 1,4-Dithiothreitol
Packaging POLYSTORMOR
Molecular Formula HSCH2(CHOH)2CH2SH
CAS 3483-12-3
Synonym DTT; Cleland's Reagent
Quantity 5g
Purity Grade Ultrapure Bioreagent
Flash Point 109.00°C
Specific Gravity 1L = 1.00kg

    For Laboratory,Research,or Manufacturing Use