Please login to your online account to display your discounted pricing

1,4-Dithiothreitol, Ultrapure Bioreagent, J.T.Baker™


High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Quantity 100g
Packaging Poly Bottle

Manufacturer: avantor performance  F78003

 View more versions of this product

Catalog No. 02002015

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 154.25
Chemical Name or Material 1,4-Dithiothreitol
Packaging Poly Bottle
Purity Grade Ultrapure Bioreagent
Molecular Formula HSCH2(CHOH)2CH2SH
CAS 3483-12-3
Synonym DTT; Cleland's Reagent
Flash Point 109.00°C
Quantity 100g
Specific Gravity 1L = 1.00kg

Specifications

Chemical Name or Material 1,4-Dithiothreitol
CAS 3483-12-3
Purity Grade Ultrapure Bioreagent
Molecular Formula HSCH2(CHOH)2CH2SH
Formula Weight 154.25
Synonym DTT; Cleland's Reagent

  • High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography.
  • For Laboratory,Research,or Manufacturing Use