Learn More
1,4-Dithioerythritol, Electrophoresis Grade, 99%, Thermo Scientific Chemicals
$81.77 - $879.40
Chemical Identifiers
CAS | 6892-68-8 |
---|---|
Molecular Formula | C4H10O2S2 |
Molecular Weight (g/mol) | 154.24 |
MDL Number | MFCD00063750 |
InChI Key | VHJLVAABSRFDPM-ZXZARUISSA-N |
Synonym | dithioerythritol, 1,4-dithioerythritol, dte, 2r,3s-1,4-dimercaptobutane-2,3-diol, erythro-1,4-dimercapto-2,3-butanediol, 2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel, 1,4-dithioerythritol dte, 2r*,3s*-1,4-dimercapto-2,3-butanediol, erythro-2,3-dihydroxy-1,4-butanedithiol, 2r,3s-1,4-disulfanylbutane-2,3-diol |
PubChem CID | 439352 |
ChEBI | CHEBI:17456 |
SMILES | O[C@@H](CS)[C@H](O)CS |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6465603
|
Thermo Scientific Chemicals
J6465603 |
1 g |
Each for $81.77
|
|
|||||
AAJ6465614
|
Thermo Scientific Chemicals
J6465614 |
25 g |
Each for $879.40
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6892-68-8 | |
154.24 | |
VHJLVAABSRFDPM-ZXZARUISSA-N | |
439352 | |
O[C@@H](CS)[C@H](O)CS |
C4H10O2S2 | |
MFCD00063750 | |
dithioerythritol, 1,4-dithioerythritol, dte, 2r,3s-1,4-dimercaptobutane-2,3-diol, erythro-1,4-dimercapto-2,3-butanediol, 2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel, 1,4-dithioerythritol dte, 2r*,3s*-1,4-dimercapto-2,3-butanediol, erythro-2,3-dihydroxy-1,4-butanedithiol, 2r,3s-1,4-disulfanylbutane-2,3-diol | |
CHEBI:17456 |
Specifications
6892-68-8 | |
White | |
C4H10O2S2 | |
1 g | |
1719756 | |
dithioerythritol, 1,4-dithioerythritol, dte, 2r,3s-1,4-dimercaptobutane-2,3-diol, erythro-1,4-dimercapto-2,3-butanediol, 2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel, 1,4-dithioerythritol dte, 2r*,3s*-1,4-dimercapto-2,3-butanediol, erythro-2,3-dihydroxy-1,4-butanedithiol, 2r,3s-1,4-disulfanylbutane-2,3-diol | |
O[C@@H](CS)[C@H](O)CS | |
154.24 | |
CHEBI:17456 | |
99% | |
Powder |
82°C to 84°C | |
Stench | |
MFCD00063750 | |
UN3335 | |
Air sensitive | |
VHJLVAABSRFDPM-ZXZARUISSA-N | |
(2R,3S)-1,4-bis(sulfanyl)butane-2,3-diol | |
439352 | |
154.25 | |
Electrophoresis | |
1,4-Dithioerythritol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H500
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 229-998-8
RTECSNumber : KF2410000
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only