Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Dipropionyloxybenzene, 98%

$79.58 - $209.34

Chemical Identifiers

Chemical Name or Material 1,4-Dipropionyloxybenzene
CAS 7402-28-0
Molecular Formula C12H14O4
Formula Weight 222.24
MDL Number MFCD00082703
Synonym Hydroquinone dipropionate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2197214

  SDS
alfa aesar™
B2197214
25g Each for $79.58

AAB2197222

  SDS
alfa aesar™
B2197222
100g Each for $209.34
Description & Specifications

Specifications

Formula Weight 222.24
Chemical Name or Material 1,4-Dipropionyloxybenzene
Assay 98%
Molecular Formula C12H14O4
MDL Number MFCD00082703
CAS 7402-28-0
Synonym Hydroquinone dipropionate
Melting Point 112° to 114°C
Quantity 25g