Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Dioxene, 98%

$132.59 - $132.59

Chemical Identifiers

Chemical Name or Material 1,4-Dioxene
CAS 543-75-9
Molecular Formula C4H6O2
Formula Weight 86.09
MDL Number MFCD00082537
UN Number UN1993
Synonym 2,3-Dihydro-1,4-dioxin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3359703

  SDS
alfa aesar™
H3359703*
1g Each for $132.59
Description & Specifications

Specifications

Formula Weight 86.09
Molecular Formula C4H6O2
MDL Number MFCD00082537
Boiling Point 94° to 96°C
Synonym 2,3-Dihydro-1,4-dioxin
UN Number UN1993
Chemical Name or Material 1,4-Dioxene
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Quantity 1g
Flash Point 1°C (34°F)
CAS 543-75-9
Assay 98%
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is ideally suited for use in closed-loop applications such as vented reactors for the manufacture of active pharmaceutical ingredients. Also find serviceable application in the synthesis of this useful heterocyclic intermediate.

Notes
Store in cool location. With adequate ventilation. Store under nitrogen. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.