missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Dioxaspiro[4.5]decan-8-ol, 98%, Thermo Scientific™

$413.94 - $413.94

Chemical Identifiers

CAS 22428-87-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.20
MDL Number MFCD00067003
InChI Key HKQTYQDNCKMNHZ-UHFFFAOYSA-N
Synonym 1,4-dioxa-spiro 4.5 decan-8-ol, 1,4-dioxaspiro 4.5 decan-8-ol, 4,4-ethylenedioxycyclohexanol, 4-hydroxycyclohexanone ethylene acetal, 4-hydroxycyclohexanone monoethylene ketal, 8-hydroxy-1,4-dioxaspiro 4.5 decane, acmc-1chv5, pubchem20110
PubChem CID 567352
IUPAC Name 1,4-dioxaspiro[4.5]decan-8-ol
SMILES OC1CCC2(CC1)OCCO2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC241010050
SDS View Documents Promotion Details
Thermo Fisher Scientific
AC241010050
5g Glass bottle
Each for $413.94
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

22428-87-1
158.20
HKQTYQDNCKMNHZ-UHFFFAOYSA-N
567352
OC1CCC2(CC1)OCCO2
C8H14O3
MFCD00067003
1,4-dioxa-spiro 4.5 decan-8-ol, 1,4-dioxaspiro 4.5 decan-8-ol, 4,4-ethylenedioxycyclohexanol, 4-hydroxycyclohexanone ethylene acetal, 4-hydroxycyclohexanone monoethylene ketal, 8-hydroxy-1,4-dioxaspiro 4.5 decane, acmc-1chv5, pubchem20110
1,4-dioxaspiro[4.5]decan-8-ol

Specifications

22428-87-1
100.0
97.5% min. (GC)
C8H14O3
Glass bottle
1,4-dioxa-spiro 4.5 decan-8-ol, 1,4-dioxaspiro 4.5 decan-8-ol, 4,4-ethylenedioxycyclohexanol, 4-hydroxycyclohexanone ethylene acetal, 4-hydroxycyclohexanone monoethylene ketal, 8-hydroxy-1,4-dioxaspiro 4.5 decane, acmc-1chv5, pubchem20110
HKQTYQDNCKMNHZ-UHFFFAOYSA-N
1,4-dioxaspiro[4.5]decan-8-ol
567352
Viscous Liquid
1, 4-Dioxaspiro[4.5]decan-8-ol
97.5
Colorless
Authentic
MFCD00067003
1.4850 to 1.487
5g
OC1CCC2(CC1)OCCO2
158.20
158.2
98%
SDS
Product Certifications

RUO – Research Use Only