missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Dioxane, puriss. p.a., ACS Reagent, Reag. Ph. Eur. reag. ISO, ≥99.5% (GC), Honeywell™ Riedel-de Haën™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5% (GC)
Supplier: Honeywell Chemicals 331476X1L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
6 to 8 (at 500g/L 20°C) | |
10°C to 12°C | |
123-91-1 | |
1.034g/mL at 25°C | |
6 x 1 L | |
12°C (53.6°F) | |
Aromatic | |
Liquid | |
MFCD00006571 | |
UN1165 | |
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
C1COCCO1 | |
88.106 | |
CHEBI:47032 | |
≥99.5% (GC) |
1,4-Dioxane | |
Reag. Ph. Eur. reag. ISO | |
Colorless | |
100°C to 102°C | |
0.22 | |
356°F | |
Glass Bottle | |
C4H8O2 | |
40mmHg (25°C), 27mmHg (20°C) | |
102551 | |
RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
1,4-dioxane | |
31275 | |
88.11g/mol | |
ACS/puriss. p.a. |
Chemical Identifiers
123-91-1 | |
88.106 | |
RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
31275 | |
1,4-dioxane |
C4H8O2 | |
MFCD00006571 | |
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
CHEBI:47032 | |
C1COCCO1 |
Safety and Handling
P210-P261-P281-P305 + P351 + P338
H225-H319-H335-H351
Danger
DOTInformation : class 3 - PG 2 - Dioxane
EINECSNumber : 204-661-8
RTECSNumber : JG8225000