Please login to your online account to display your discounted pricing

1,4-Dioxane-d8, for NMR, 99% atom D, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 4-Dioxane-d8
Name Note 99 atom % D
CAS 17647-74-4
Purity Grade For NMR
Molecular Formula C4 D8 O2
Formula Weight 96.17g/mol
MDL Number MFCD00044239
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC181450050

  SDS
thermo fisher scientific
181450050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Deuterium (D) 98.5% min.
Packaging Glass bottle
Density 1.1200g/mL
Quantity 5g
Color Clear colorless
Chemical Name or Material 1, 4-Dioxane-d8
Name Note 99 atom % D
CAS 17647-74-4
Purity Grade For NMR
Physical Form Clear colorless liquid
Molecular Formula C4 D8 O2
Formula Weight 96.17g/mol
MDL Number MFCD00044239
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
May cause respiratory irritation.
Suspected of causing cancer.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 241-628-7
TSCA TSCA

For NMR

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
May cause respiratory irritation.
Suspected of causing cancer.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.