Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Dioxane-d8, 99% (Isotopic)

$65.69 - $65.69

Chemical Identifiers

Chemical Name or Material 1,4-Dioxane-d8
CAS 17647-74-4
Name Note Isotopic
Formula Weight 96.16
MDL Number MFCD00044239
UN Number UN1165
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3651603

  SDS
alfa aesar™
3651603
1g Each for $65.69
Description & Specifications

Specifications

Formula Weight 96.16
Chemical Name or Material 1,4-Dioxane-d8
Sensitivity Hygroscopic
Density 1.13g/mL
Name Note Isotopic
Boiling Point 101°C
Melting Point 12°C
Physical Form Liquid
Assay 99%
Quantity 1g
MDL Number MFCD00044239
CAS 17647-74-4
UN Number UN1165
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H351-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIOXANE
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H351-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.