missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Dioxane, anhydrous, 99.8%, Alfa Aesar™

Manufacturer:  Alfa Aesar 43821K2

 View more versions of this product

Catalog No. AA43821K2



Specifications

Specifications

1, 4-Dioxane, anhydrous
Colorless liquid
C4H8O2
1165
1, 4-Dioxane, anhydrous
C1COCCO1
88.106
CHEBI:47032
Liquid
Ether like
12°C (53.6°F)
Colorless
1L
123-91-1
99.8%
MFCD00006571
1,4-diethylene dioxide
RYHBNJHYFVUHQT-UHFFFAOYSA-N
1,4-dioxane
31275
88.11
375°C (707°F)
41 hPa (31mm Hg) at 20°C
100°C to 102°C
11.8°C

Chemical Identifiers

123-91-1
88.106
RYHBNJHYFVUHQT-UHFFFAOYSA-N
31275
1,4-dioxane
C4H8O2
MFCD00006571
1,4-diethylene dioxide
CHEBI:47032
C1COCCO1
Safety and Handling

Safety and Handling

H225-H351-H319-H335: Highly flammable liquid and vapour. Suspected of causing cancer. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 204-661-8

Documents