Learn More
1,4-Dioxane, 99+%, extra pure, stabilized, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 117110010
Specifications
1, 4-Dioxane | |
123-91-1 | |
100.0 | |
C4H8O2 | |
19, 3 | |
15, 3328 | |
RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
1,4-dioxane | |
31275 | |
88.11 | |
≥99% | |
1.0340g/mL | |
0.005% max. (as H2O2) | |
0.01% max. | |
1.034 | |
0.1% max. | |
1.32 mPa.s (20°C) | |
Colorless | |
1L |
99+% | |
99.0 | |
99+% | |
MFCD00006571 | |
01,333 | |
1,4-diethylene dioxide, 1,4-dioxacyclohexane, di ethylene oxide, diethylene dioxide, diethylene ether, dioxan, dioxane, dioxanne, p-dioxane, tetrahydro-p-dioxin | |
C1COCCO1 | |
88.106 | |
CHEBI:47032 | |
Liquid | |
Extra Pure | |
Glass bottle | |
1.4205 to 1.4220 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents | |
5 to 30ppm BHT | |
12°C | |
101.0°C | |
12.0°C |
Chemical Identifiers
123-91-1 | |
88.106 | |
RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
31275 | |
1,4-dioxane |
C4H8O2 | |
MFCD00006571 | |
1,4-diethylene dioxide, 1,4-dioxacyclohexane, di ethylene oxide, diethylene dioxide, diethylene ether, dioxan, dioxane, dioxanne, p-dioxane, tetrahydro-p-dioxin | |
CHEBI:47032 | |
C1COCCO1 |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 204-661-8
RTECSNumber : JG8225000
TSCA : TSCA