missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Dioxane, 99+%, ACS reagent, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 408825000

 View more versions of this product

Catalog No. AC408825000


 

Specifications Description & Specifications

Specifications

≥99.0%
≤11.0°C
Liquid
Clear
1.0340g/mL
01,333
100.0
0.005% max.
0.01% max.
0.005% max. (H2O2)
0.05% max. (Coulometric)
1, 4-Dioxane
123-91-1
ACS Reagent Grade
88.11
500mL
EcoSafPak™ Glass Bottle
15, 3328
19, 3
99.0
0.0016 meq/g max.
MFCD00006571
20 to 30ppm (ca.) BHT
Diox
C4H8O2
99+%

Chemical Identifiers

123-91-1
100.0
C4H8O2
MFCD00006571
1, 4-Dioxane
ACS Reagent Grade
99.0
99+%
88.11
Diox
≥99.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.

Emergency Overview
Flammable liquid and vapor. May form explosive peroxides. Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. Inhalation may cause central nervous system effects. Repeated exposure may cause skin dryness or cracking. Hygroscopic.
NFPA
Health: 2
Flammability: 3
Instability: 1

SIGNAL WORD: DANGER!

EINECSNumber : 204-661-8

RTECSNumber : JG8225000

TSCA : TSCA