Learn More
1,4-Dioxane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
1, 4-Dioxane, 99.5%, C4H8O2, CAS Number-123-91-1, diethylene dioxide, 1,4-dioxacyclohexane, 1,4-diethylene dioxide, dioxanne, dioxan, di ethylene oxide, diethylene ether, tetrahydro-p-dioxin, p-dioxane, dioxane, 1L, 0.002 % max., 19, 3, 101.0 deg.C, 100.0, 99.4, CHEBI:47032, Yellow to Brown
Supplier: Thermo Scientific Chemicals 364340010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 4-Dioxane | |
Over Molecular Sieves | |
Yellow to Brown | |
100.0 | |
101.0°C | |
12°C | |
AcroSeal™ Glass Bottle | |
1.4205 to 1.4220 | |
1.034 | |
MFCD00006571 | |
1165 | |
19, 3 | |
15, 3328 | |
RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
1,4-dioxane | |
31275 | |
88.11 | |
Extra Dry over Molecular Sieve |
12.0°C | |
123-91-1 | |
99.4 | |
1.0340g/mL | |
1 L | |
Authentic | |
Liquid | |
99.5% | |
C4H8O2 | |
1.32 mPa.s (20°C) | |
0.005% Max. | |
01,333 | |
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
C1COCCO1 | |
88.106 | |
CHEBI:47032 | |
99.5% |
Chemical Identifiers
123-91-1 | |
88.106 | |
RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
31275 | |
1,4-dioxane |
C4H8O2 | |
MFCD00006571 | |
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
CHEBI:47032 | |
C1COCCO1 |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
Emergency Overview
Flammable liquid and vapor. May form explosive peroxides. Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes
SIGNAL WORD: DANGER!
RUO – Research Use Only