missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Dioxane, 99.5%, for analysis, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC270490250

 View more versions of this product

Catalog No. AC270490250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1,4-Dioxane
123-91-1
100.0
C4H8O2
19, 3
15, 3328
Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents
C1COCCO1
88.106
31275
88.11
99.5%
0.1ppm max.
1.0340g/mL
0.1ppm max.
0.005% max. (as H2O2)
Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents
25ppm (ca.) BHT
12°C
101.0°C
12.0°C
Stabilized
99.4
99.5%
MFCD00006571
01,333
1,4-diethylene dioxide, 1,4-dioxacyclohexane, di ethylene oxide, diethylene dioxide, diethylene ether, dioxan, dioxane, dioxanne, p-dioxane, tetrahydro-p-dioxin
RYHBNJHYFVUHQT-UHFFFAOYSA-N
1,4-dioxane
0.1ppm max.
CHEBI:47032
Liquid
Analysis
0.1ppm max.
0.001% max.
Metal Drum
1.4205 to 1.4220
1.034
0.1% max.
1.32 mPa.s (20°C)
Undesignated
25L

Chemical Identifiers

123-91-1
88.106
RYHBNJHYFVUHQT-UHFFFAOYSA-N
31275
1,4-dioxane
C4H8O2
MFCD00006571
1,4-diethylene dioxide, 1,4-dioxacyclohexane, di ethylene oxide, diethylene dioxide, diethylene ether, dioxan, dioxane, dioxanne, p-dioxane, tetrahydro-p-dioxin
CHEBI:47032
C1COCCO1
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents