Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Dioxane-2,3-diol, 97%

$55.89 - $226.01

Chemical Identifiers

Chemical Name or Material 1,4-Dioxane-2,3-diol
CAS 4845-50-5
Molecular Formula C4H8O4
Formula Weight 120.11
MDL Number MFCD00006572
Beilstein 104209
Synonym 2,3-Dihydroxy-1,4-dioxane; Glyoxal monoethylene acetal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1921006

  SDS
alfa aesar™
L1921006
5g Each for $55.89

AAL1921014

  SDS
alfa aesar™
L1921014
25g Each for $226.01
Description & Specifications

Specifications

Formula Weight 120.11
Assay 97%
Molecular Formula C4H8O4
MDL Number MFCD00006572
CAS 4845-50-5
Beilstein 104209
Synonym 2,3-Dihydroxy-1,4-dioxane; Glyoxal monoethylene acetal
Health Hazard 1 Warning
Melting Point 100° to 102°C
Chemical Name or Material 1,4-Dioxane-2,3-diol
TSCA TSCA
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Quantity 5g

Applications
1,4-Dioxane-2,3-diol is used as a pharmaceutical intermediate and an intermediate in organic synthesis.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.