Please login to your online account to display your discounted pricing

1,4-Dioxa-8-azaspiro[4.5]decane, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 4-Dioxa-8-azaspiro[4.5]decane
Name Note 98%
CAS 177-11-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H13NO2
Formula Weight 143.19
MDL Number MFCD00005976
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC176370500

  SDS
fisher scientific acros
176370500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC176370100

  SDS
fisher scientific acros
176370100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 143.19
Quantity 50g
Color Clear colorless to yellow
EINECS Number 205-868-6
TSCA TSCA
Density 1.1100g/mL
CAS Max % 100.0
Chemical Name or Material 1, 4-Dioxa-8-azaspiro[4.5]decane
Packaging Glass bottle
Refractive Index 1.4809 to 1.4829 (20°C, 589nm)
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H13NO2
MDL Number MFCD00005976
CAS 177-11-7
Synonym 4-Piperidone ethylene ketal
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.