missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-dimethylpiperazine-2-carboxylic acid dihydrochloride, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC13851CB
Specifications
1, 4-dimethylpiperazine-2-carboxylic acid dihydrochloride | |
Amber Glass Bottle | |
250mg | |
UPJQXMOMHHDOSZ-UHFFFAOYSA-N | |
1,4-dimethylpiperazine-2-carboxylic acid;dihydrochloride | |
45594293 | |
95% |
1174064-55-1 | |
C7H16Cl2N2O2 | |
1,4-dimethylpiperazine-2-carboxylic acid dihydrochloride, 1,4-dimethylpiperazine-2-carboxylic acid,dihydrochloride, 1,4-dimethylpiperazine-2-carboxylic acid, chloride, chloride, 1,4-dimethylpiperazine-2-carboxylic acid-hydrogen chloride 1/2 | |
CN1CCN(C(C1)C(=O)O)C.Cl.Cl | |
231.117 | |
231.12 |
Chemical Identifiers
1174064-55-1 | |
231.117 | |
1,4-dimethylpiperazine-2-carboxylic acid dihydrochloride, 1,4-dimethylpiperazine-2-carboxylic acid,dihydrochloride, 1,4-dimethylpiperazine-2-carboxylic acid, chloride, chloride, 1,4-dimethylpiperazine-2-carboxylic acid-hydrogen chloride 1/2 | |
1,4-dimethylpiperazine-2-carboxylic acid;dihydrochloride |
C7H16Cl2N2O2 | |
UPJQXMOMHHDOSZ-UHFFFAOYSA-N | |
45594293 | |
CN1CCN(C(C1)C(=O)O)C.Cl.Cl |