missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Dimethyl-2-iodobenzene 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC393250050

 View more versions of this product

Catalog No. AC393250050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

1, 4-Dimethyl-2-iodobenzene
5g
97.5
1.617g/mL
Glass bottle
Liquid
98%
IC6H3(CH3)2
MFCD00013708
Clear yellow
1122-42-5
100.0
Authentic
1.5960 to 1.5980 (20°C, 589nm)
≥97.5%
C8H9I
232.06

Chemical Identifiers

1122-42-5
100.0
C8H9I
232.06
1, 4-Dimethyl-2-iodobenzene
97.5
98%
IC6H3(CH3)2
MFCD00013708
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.