Please login to your online account to display your discounted pricing

(1,4-Dimethyl-1,2,3,4-tetrahydroquinoxalin-6-yl)methylamine, ≥90%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC37113CB

 View more versions of this product

Catalog No. CC37113CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 191.28
Quantity 250mg
Assay 90%
CAS Max % 100.0
Molecular Formula C11H17N3
MDL Number MFCD06797483
CAS Min % 89.5
CAS 850375-15-4
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material (1,4-Dimethyl-1,2,3,4-tetrahydroquinoxalin-6-yl)methylamine
Assay Percent Range 90%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

Specifications

Chemical Name or Material (1,4-Dimethyl-1,2,3,4-tetrahydroquinoxalin-6-yl)methylamine
CAS 850375-15-4
CAS Min % 89.5
CAS Max % 100.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C11H17N3
Formula Weight 191.28
MDL Number MFCD06797483
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

SDS & Safety Documents