Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Dimethoxynaphthalene, 96%

$100.96 - $1,488.68

Chemical Identifiers

Chemical Name or Material 1,4-Dimethoxynaphthalene
CAS 10075-62-4
Molecular Formula C12H12O2
Formula Weight 188.23
MDL Number MFCD00052378
Beilstein 2048082
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1409503

 
alfa aesar™
A1409503
1g Each for $100.96

AAA1409506

 
alfa aesar™
A1409506
5g Each for $358.61

AAA1409514

 
alfa aesar™
A1409514
25g Each for $1,488.68
Description & Specifications

Specifications

Formula Weight 188.23
Melting Point 84° to 85°C
MDL Number MFCD00052378
Assay 96%
CAS 10075-62-4
Beilstein 2048082
Chemical Name or Material 1,4-Dimethoxynaphthalene
Molecular Formula C12H12O2
Quantity 1g

Applications
1,4-Dimethoxynaphthalene may be used as a reference molecule during the synthesis, separation and analysis of derivatized naphthalenes.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.