Learn More
1,4-Dimethoxybenzene, 99+%, Thermo Scientific Chemicals
$52.99 - $203.89
Chemical Identifiers
CAS | 150-78-7 |
---|---|
Molecular Formula | C8H10O2 |
Molecular Weight (g/mol) | 138.17 |
MDL Number | MFCD00008401 |
InChI Key | OHBQPCCCRFSCAX-UHFFFAOYSA-N |
Synonym | p-dimethoxybenzene, p-methoxyanisole, hydroquinone dimethyl ether, benzene, 1,4-dimethoxy, quinol dimethyl ether, benzene, p-dimethoxy, dimethylhydroquinone ether, dimethyl ether hydroquinone, 1,4-dimethoxybenzol, usaf an-9 |
PubChem CID | 9016 |
IUPAC Name | 1,4-dimethoxybenzene |
SMILES | COC1=CC=C(OC)C=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC115411000
|
Thermo Scientific Chemicals
115411000 |
100 g | Glass bottle |
Each for $52.99
|
|
||||
AC115415000
|
Thermo Scientific Chemicals
115415000 |
500 g | Glass bottle |
Each for $118.58
|
|
||||
AC115410010
|
Thermo Scientific Chemicals
115410010 |
1 kg | Glass bottle |
Each for $203.89
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
150-78-7 | |
138.17 | |
OHBQPCCCRFSCAX-UHFFFAOYSA-N | |
9016 | |
COC1=CC=C(OC)C=C1 |
C8H10O2 | |
MFCD00008401 | |
p-dimethoxybenzene, p-methoxyanisole, hydroquinone dimethyl ether, benzene, 1,4-dimethoxy, quinol dimethyl ether, benzene, p-dimethoxy, dimethylhydroquinone ether, dimethyl ether hydroquinone, 1,4-dimethoxybenzol, usaf an-9 | |
1,4-dimethoxybenzene |
Specifications
150-78-7 | |
100.0 | |
White | |
213°C | |
Authentic | |
Glass bottle | |
C6H4(OCH3)2 | |
100 g | |
1.05 | |
Solubility in water: 0.8g/L (20 c). Other solubilities: soluble in benzene,alcohol,ethylacetate and,acetone,1g/10mL dioxane | |
COC1=CC=C(OC)C=C1 | |
138.17 | |
1.04 mPa.s (65°C) | |
99+% | |
1, 4-Dimethoxybenzene, 99+% |
99.0 | |
54°C to 58°C | |
1.0500g/mL | |
98°C | |
99% min. (GC) | |
C8H10O2 | |
MFCD00008401 | |
06, 843 | |
p-dimethoxybenzene, p-methoxyanisole, hydroquinone dimethyl ether, benzene, 1,4-dimethoxy, quinol dimethyl ether, benzene, p-dimethoxy, dimethylhydroquinone ether, dimethyl ether hydroquinone, 1,4-dimethoxybenzol, usaf an-9 | |
OHBQPCCCRFSCAX-UHFFFAOYSA-N | |
1,4-dimethoxybenzene | |
9016 | |
138.17 | |
Crystalline Powder, Crystals or Flakes |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 205-771-9
RTECSNumber : CZ6650000
TSCA : TSCA
RUO – Research Use Only