missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Diiodobenzene, 98%

Manufacturer:  Alfa Aesar™ A1533514

 View more versions of this product

Catalog No. AAA1533514



Description

Description

Applications
1,4-Diiodobenzene is used in Suzuki reaction. It is used as a precursor in the preparation of martinellic acid and 1,4-bis(p-R-phenylethynyl)benzenes. Further, it is employed in the preparation of 1,4-diiodo-2,5-didodecylbenzene , which is a starting reagent for the synthesis of oligo(1,4-phenylene ethynylene). It is also used in human and animal nutrition products including antiseptics and disinfectants. In addition to this, it used in polarizing films for liquid crystal display chemicals.

Solubility
Soluble in ether and ethanol. Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications

Specifications

1,4-Diiodobenzene
128°C to 132°C
624-38-4
MFCD00001054
p-diiodobenzene, benzene, 1,4-diiodo, p-benzene diiodide, benzene, p-diiodo, unii-i9xcf83vav, i9xcf83vav, 1,4-diiodbenzene, 1,4-diidobenzene, 1,4 diiodobenzene, 1,4-diiodiobenzene
C1=CC(=CC=C1I)I
329.907
329.91
285°C
25g
C6H4I2
1904546
LFMWZTSOMGDDJU-UHFFFAOYSA-N
1,4-diiodobenzene
12208
98%

Chemical Identifiers

624-38-4
329.907
LFMWZTSOMGDDJU-UHFFFAOYSA-N
12208
C1=CC(=CC=C1I)I
C6H4I2
MFCD00001054
p-diiodobenzene, benzene, 1,4-diiodo, p-benzene diiodide, benzene, p-diiodo, unii-i9xcf83vav, i9xcf83vav, 1,4-diiodbenzene, 1,4-diidobenzene, 1,4 diiodobenzene, 1,4-diiodiobenzene
1,4-diiodobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents