missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Dihydroxyanthraquinone, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 132180050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 4-Dihydroxyanthraquinone | |
81-64-1 | |
100.0 | |
Brown-Red to Orange | |
222°C | |
96% | |
C14H8O4 | |
5 g | |
15, 8180 | |
Solubility in water: <1g/L (20°C). Other solubilities: soluble in hot water,soluble in alcohol,ether,benzene,sulfuric,acid,potassium chloride,chloroform,acetone,,aqueous alkali and other organic solvents | |
C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)O | |
240.21 | |
CHEBI:37487 | |
96% |
96% | |
95.0 | |
191.0°C to 193.0°C | |
450.0°C | |
Authentic | |
Glass bottle | |
MFCD00001209 | |
08, 450 | |
quinizarin, 1,4-dihydroxyanthraquinone, quinizarine, chinizarin, solvent orange 86, 1,4-dihydroxyanthrachinon, smoke orange r, 1,4-dihydroxy-9,10-anthraquinone, macrolex orange gg, 9,10-anthracenedione, 1,4-dihydroxy | |
GUEIZVNYDFNHJU-UHFFFAOYSA-N | |
1,4-dihydroxyanthracene-9,10-dione | |
6688 | |
240.21 | |
Crystalline Powder |
Chemical Identifiers
81-64-1 | |
240.21 | |
GUEIZVNYDFNHJU-UHFFFAOYSA-N | |
6688 | |
1,4-dihydroxyanthracene-9,10-dione |
C14H8O4 | |
MFCD00001209 | |
quinizarin, 1,4-dihydroxyanthraquinone, quinizarine, chinizarin, solvent orange 86, 1,4-dihydroxyanthrachinon, smoke orange r, 1,4-dihydroxy-9,10-anthraquinone, macrolex orange gg, 9,10-anthracenedione, 1,4-dihydroxy | |
CHEBI:37487 | |
C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)O |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
GHS Signal Word: Warning
EINECSNumber : 201-368-7
RTECSNumber : CB6600000
TSCA : TSCA
RUO – Research Use Only