Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Diformylpiperazine, 98+%

$54.78 - $791.19

Chemical Identifiers

Chemical Name or Material 1,4-Diformylpiperazine
CAS 4164-39-0
Molecular Formula C6H10N2O2
Formula Weight 142.16
MDL Number MFCD00006153
Beilstein 121175
Synonym 1,4-Piperazinedicarboxaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1809714

  SDS
alfa aesar™
A1809714
25g Each for $54.78

AAA1809736

  SDS
alfa aesar™
A1809736
500g Each for $791.19

AAA1809722

  SDS
alfa aesar™
A1809722
100g Each for $181.64
Description & Specifications

Specifications

Formula Weight 142.16
Chemical Name or Material 1,4-Diformylpiperazine
TSCA TSCA
Assay ≥98%
Molecular Formula C6H10N2O2
MDL Number MFCD00006153
CAS 4164-39-0
Beilstein 121175
Synonym 1,4-Piperazinedicarboxaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Odor Odorless
Boiling Point 154°C (0.25mmHg)
Melting Point 126° to 129°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.