missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Difluorobenzene, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 114611000

 View more versions of this product

Catalog No. AC114611000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1, 4-Difluorobenzene
2°C
Glass bottle
1.17
Colorless to Yellow
100mL
540-36-3
100.0
C6H4F2
MFCD00000344
15, 3167
QUGUFLJIAFISSW-UHFFFAOYSA-N
1,4-difluorobenzene
10892
114.09
99+%
1.1700g/mL
Authentic
1.4400 to 1.442
88°C to 89°C
-13°C
99+%
99.0
99+%
C6H4F2
05, 199
1,4-difluorbenzol, 2,5-difluorobenzene, benzene, 1,4-difluoro, benzene, p-difluoro, benzene,4-difluoro, p-difluorobenzene, para-difluorobenzene, pubchem2309, unii-y9fq2ef0z6, y9fq2ef0z6
FC1=CC=C(F)C=C1
114.10
CHEBI:38585
Liquid

Chemical Identifiers

540-36-3
114.10
QUGUFLJIAFISSW-UHFFFAOYSA-N
10892
1,4-difluorobenzene
C6H4F2
MFCD00000344
1,4-difluorbenzol, 2,5-difluorobenzene, benzene, 1,4-difluoro, benzene, p-difluoro, benzene,4-difluoro, p-difluorobenzene, para-difluorobenzene, pubchem2309, unii-y9fq2ef0z6, y9fq2ef0z6
CHEBI:38585
FC1=CC=C(F)C=C1
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-742-9

RTECSNumber : CZ5658000

TSCA : TSCA

SDS Documents
Product Certifications